missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium, Technical, Metal Sticks In Liquid Paraffin, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Amber glass bottle
Specifications
Color | Gray |
CAS | 7440-23-5 |
Melting Point | 98°C |
Boiling Point | 883°C |
Molecular Formula | Na |
MDL Number | MFCD00085307 |
Quantity | 100 g |
Synonym | natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot |
InChI Key | KEAYESYHFKHZAL-UHFFFAOYSA-N |
SMILES | [Na] |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Sodium, under mineral oil
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'