missing translation for 'onlineSavingsMsg'
Learn More
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals Product Code.: 11448797

Sodium tert-butoxide, 97%, Thermo Scientific Chemicals

Product Code. 11448797
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11448797

Brand: Thermo Scientific Alfa Aesar A19942.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.105
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxideShow MoreShow Less
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Density 0.785
Odor Odorless
Linear Formula (CH3)3CONa
Quantity 100 g
UN Number UN3206
Beilstein 3654215
Sensitivity Air and moisture sensitive
Formula Weight 96.11
Percent Purity 97%
Chemical Name or Material Sodium tert-butoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460362 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA19942.0E,A19942.22,A19942.36
Lot NumberA0446795 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberA19942.0E,A19942.22,A19942.36
Lot Number10244775 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberA19942.0E,A19942.22,A19942.36
Lot NumberW01I010 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog NumberA19942.0E,A19942.22,A19942.36
Lot Number10242106 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog NumberA19942.0E,A19942.22,A19942.36
compliance-icons compliance-icons
Product Identifier
  • Sodium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H228-Flammable solid.
  • H251-Self-heating: may catch fire.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium 2-methylpropan-2-olate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.