missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-butoxide, 99%, pure

Product Code. 10723924 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10723924

Brand: Thermo Scientific Acros 252555000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-48-5 | C4H9NaO | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.1
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Melting Point 180.0°C
Flash Point 12°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NaOC(CH3)3
Quantity 500 g
Fieser 16,55
Solubility Information Solubility in water: reacts.
Formula Weight 96.1
Percent Purity 99%
Grade Pure
Chemical Name or Material Sodium tert-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H251-Self-heating: may catch fire.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.