missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-pentoxide, 2.5M (30 wt%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

£56.00 - £259.00

Chemical Identifiers

CAS 14593-46-5, 109-99-9
Molecular Formula C5H11NaO
Molecular Weight (g/mol) 110.13
MDL Number MFCD00064245
InChI Key CGRKYEALWSRNJS-UHFFFAOYSA-N
Synonym sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
PubChem CID 2733478
IUPAC Name sodium;2-methylbutan-2-olate
SMILES CCC(C)(C)[O-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10722084
View Documents
Thermo Scientific Acros
390151000
100 mL AcroSeal™ Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10370624
View Documents
Thermo Scientific Acros
390158000
800 mL AcroSeal™ Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14593-46-5, 109-99-9
110.13
CGRKYEALWSRNJS-UHFFFAOYSA-N
2733478
CCC(C)(C)[O-].[Na+]
C5H11NaO
MFCD00064245
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
sodium;2-methylbutan-2-olate

Specifications

66°C (1013.0 hPa)
14593-46-5, 109-99-9
CH3CH2C(CH3)2ONa
−21°C
Solubility in water: reacts
CCC(C)(C)[O-].[Na+]
110.13
110.13
Yellow to light brown solution
0.9200g/mL
< 5°C
C5H11NaO
MFCD00064245
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
CGRKYEALWSRNJS-UHFFFAOYSA-N
sodium;2-methylbutan-2-olate
2733478
29 to 31%
0.92
Sodium tert-pentoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 238-639-4

Product Identifier
  • Sodium tert-pentoxide, 30% solution in THF

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only