missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis

Specifications
Specifications
Melting Point | 75.0°C |
Color | White |
pH | 9 to 9.5 (5% soln. at 20°C) |
Physical Form | Fine Crystalline Powder |
Quantity | 1 kg |
Assay Percent Range | 99.5% |
Linear Formula | Na2B4O7·10H2O |
Fieser | 04,461 |
Merck Index | 15,8726 |
Solubility Information | Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol |
Show More |
Safety and Handling


Product Identifier
- Sodium tetraborate decahydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction