missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Honeywell Fluka™

Product Code. 15608160 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
Unit Size:
2.5kg
25g
500g
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15608160

Brand: Honeywell-Fluka S9640500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 500 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium tetraborate decahydrate
compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.