missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Honeywell Fluka™

Product Code. 15688150 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
Unit Size:
2.5kg
25g
500g
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15688150

Brand: Honeywell-Fluka S964025G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 25 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium tetraborate decahydrate
compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.