missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™
Click to view available options
Quantity:
1 g
5 g
Description
Sodium tetrabromoaurate(III) hydrate participates in the fabricated of core/alloy nanoparticles via the layer-by-layer hydrothermal route ans its growth characteristics and optical properties study.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in water |
IUPAC Name | sodium;tetrabromogold(1-);hydrate |
Formula Weight | 539.59 (Anhydrous) |
Odor | Odorless |
Appearance | Crystalline powder |
Chemical Name or Material | Sodium tetrabromoaurate(III) hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'