missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroaluminate, Thermo Scientific Chemicals

Product Code. 11318037
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-16-9
Molecular Formula AlCl4Na
Molecular Weight (g/mol) 191.771
MDL Number MFCD00050361
InChI Key CMLRNXGMTZKKSR-UHFFFAOYSA-J
Synonym sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis
PubChem CID 16699350
IUPAC Name sodium;tetrachloroalumanuide
SMILES [Na+].[Al-](Cl)(Cl)(Cl)Cl

Specifications

Melting Point 185°C
Color Yellow
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 191.78
Sensitivity Moisture sensitive
Appearance Pale yellow powder
Density 2.01
Chemical Name or Material Sodium tetrachloroaluminate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'