Learn More
Sodium tetrachloroplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 42.4% min, Thermo Scientific Chemicals
£127.00 - £523.00
Chemical Identifiers
CAS | 207683-21-4 |
---|---|
Molecular Formula | Cl4Na2Pt |
Molecular Weight (g/mol) | 382.87 |
MDL Number | MFCD00150188 |
InChI Key | DOAIHRYZKUCMDX-UHFFFAOYSA-J |
Synonym | sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1 |
PubChem CID | 56846332 |
IUPAC Name | disodium;tetrachloroplatinum(2-);hydrate |
SMILES | [Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316548
|
Thermo Scientific Alfa Aesar
012168.03 |
1 g |
£127.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306548
|
Thermo Scientific Alfa Aesar
012168.06 |
5 g |
£523.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tetrachloroplatinate(II) hydrate is used as chemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207683-21-4 | |
382.87 | |
DOAIHRYZKUCMDX-UHFFFAOYSA-J | |
56846332 | |
[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--] |
Cl4Na2Pt | |
MFCD00150188 | |
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1 | |
disodium;tetrachloroplatinum(2-);hydrate |
Specifications
100°C | |
Crystalline | |
MFCD00150188 | |
Soluble in water. | |
[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--] | |
382.87 | |
382.88 (Anhydrous) | |
Premion™ | |
(metals basis) |
207683-21-4 | |
Cl4Na2Pt | |
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1 | |
DOAIHRYZKUCMDX-UHFFFAOYSA-J | |
disodium;tetrachloroplatinum(2-);hydrate | |
56846332 | |
99.95% | |
Pt 42.4% min. | |
Sodium tetrachloroplatinate(II) hydrate |
Safety and Handling
GHS H Statement
H300-H334-H318-H315-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302+H312+H332-H315-H317-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 233-051-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only