missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrafluoroborate, 97+%, extra pure, Thermo Scientific Chemicals

£20.10 - £188.94

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
SMILES [Na+].F[B-](F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10457940
View Documents
Thermo Scientific Acros
192120250
25 g Plastic bottle
£20.10
25g
Please to purchase this item. Need a web account? Register with us today!
10062610
View Documents
Thermo Scientific Acros
192125000
500 g Plastic bottle
£42.81
500g
Please to purchase this item. Need a web account? Register with us today!
10165103
View Documents
Thermo Scientific Acros
192120025
2.5 kg Plastic bottle
£188.94
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

13755-29-8
109.79
KGJZTOFHXCFQIV-UHFFFAOYSA-N
4343483
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
[Na+].F[B-](F)(F)F

Specifications

384°C
Brown to White
97+%
NaBF4
15, 8753
Solubility in water: 973g/L (20°C). Other solubilities: sparingly soluble in alcohol
[Na+].F[B-](F)(F)F
109.79
109.79
Extra Pure
Sodium tetrafluoroborate, 98%
13755-29-8
Powder, Needles or Crystals
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
KGJZTOFHXCFQIV-UHFFFAOYSA-N
sodium tetrafluoroboranuide
4343483
≥97%
2.47
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 237-340-6

missing translation for 'rtecsNumber' : ED2975000

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium tetrafluoroborate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only