missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium tetrathionate dihydrate

Product Code. 15783487
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-29-4
Molecular Formula H4Na2O8S4
Molecular Weight (g/mol) 306.24
MDL Number MFCD00150868
InChI Key HAEPBEMBOAIUPN-UHFFFAOYSA-L
Synonym sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2
PubChem CID 203055
IUPAC Name disodium dihydrate (sulfonatodisulfanyl)sulfonate
SMILES O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O

Specifications

Quantity 25 g
Solubility Information Solubility in water: 30.6g/L.
Formula Weight 306.27
Chemical Name or Material Sodium tetrathionate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'