missing translation for 'onlineSavingsMsg'
Learn More
Sodium, Thermo Scientific Chemicals Product Code.: 10141052

Sodium, Thermo Scientific Chemicals

Product Code. 10141052
250 g, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10141052

Brand: Thermo Scientific Acros 370212500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Color Gray or White
Melting Point 98.0°C
Boiling Point 883.0°C
Linear Formula Na
Quantity 250 g
Fieser 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303
Formula Weight 22.99
Percent Purity ≥99.8%
Packaging Glass bottle
Form Stick
Chemical Name or Material Sodium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463530 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number370210010,370212500
Lot NumberA0463530 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number370210010,370212500
Lot NumberA0451205 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number370210010,370212500
Lot NumberA0443913 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number370210010,370212500
Lot NumberA0398683 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number370210010,370212500
compliance-icons compliance-icons
Product Identifier
  • Sodium, dry stick
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.