missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiocyanate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
Solubility
Soluble in water (1250g/L), alcohol, and acetone.
Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
Solubility
Soluble in water (1250g/L), alcohol, and acetone.
Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
Specifications
Specifications
Melting Point | 287°C |
Density | 1.74 |
pH | 6 to 8 |
Odor | Odorless |
Quantity | 250 g |
Beilstein | 3594965 |
Sensitivity | Hygroscopic; Light sensitive |
Merck Index | 14,9327 |
Solubility Information | Soluble in water (1250g/L),alcohol,and acetone. |
Formula Weight | 81.07 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'