missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiocyanate, 98+%, for analysis
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 287.0°C |
| Quantity | 1 kg |
| Assay Percent Range | 98% min. (Argentometry) |
| Linear Formula | NaSCN |
| Merck Index | 15,948 |
| Solubility Information | Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone |
| Formula Weight | 81.07 |
| Percent Purity | ≥98% |
| Grade | Analytical |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
Product Identifier
- Sodium thiocyanate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- P233-Keep container tightly closed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Sodium thiocyanate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction