missing translation for 'onlineSavingsMsg'
Learn More
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals Product Code.: 10043710

Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals

Product Code. 10043710
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10043710

Brand: Thermo Scientific Acros 305710050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.09
MDL Number MFCD00174316
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
PubChem CID 4378561
SMILES C[S-].[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 94 to 106% (Iodimetry)
Packaging Glass bottle
Linear Formula CH3SNa
Quantity 5 g
Synonym sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium
Solubility Information Solubility in water: freely soluble.
IUPAC Name sodium;methanethiolate
Formula Weight 70.09
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium thiomethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468274 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number305710050,305710250,305710010,305711000
Lot NumberA0469100 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number305710050,305710250,305710010,305711000
Lot NumberA0451829 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number305710050,305710250,305710010,305711000
Lot NumberA0463489 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number305710050,305710250,305710010,305711000
Lot NumberA0451829 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number305710050,305710250,305710010,305711000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium thiomethoxide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.