missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiophenolate, 90%, Thermo Scientific Chemicals

Product Code. 11985751
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-69-8
Molecular Formula C6H5NaS
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066460
InChI Key RZWQDAUIUBVCDD-UHFFFAOYSA-M
Synonym sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
PubChem CID 4110039
IUPAC Name sodium;benzenethiolate
SMILES [Na+].[S-]C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Beige to White
Physical Form Powder
Quantity 10 g
Assay Percent Range 88% min. (Iodimetry)
Linear Formula C6H5SNa
Solubility Information Solubility in water: soluble
Formula Weight 132.16
Percent Purity 90%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sodium thiophenolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium thiophenolate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.