Learn More
Sodium thiophosphate hydrate, Thermo Scientific Chemicals
Description
Sodium thiophosphate hydrate is used in thiosulfate-assisted synthesis of a variety of diamine-CuCN complexes. In analytical chemistry, it is used as a titrant in iodometry. It finds application as an anionic surfactant in pharmaceutical preparations to assist in dispersion and an antidote to cyanide poisoning. It is utilized for the dechlorination of tap water as well as to maintain the chlorine levels in swimming pools and aquaria. It plays an important role in gold extraction. It is also used in the alkylation reaction to prepare Bunte salts viz. S-alkylthiosulfonates, which finds application in the synthesis of thioglycolic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 53°C to 58°C |
Color | Green to White |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Merck Index | 14,8693 |
Solubility Information | Soluble in water. |
Formula Weight | 179.93 (Anhydrous) |
Chemical Name or Material | Sodium thiophosphate hydrate |
RUO – Research Use Only