missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, 99+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
Melting Point | 48.5°C |
Color | Colorless to White |
pH | 6 to 8.4 (10% soln. at 20°C) |
Physical Form | Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99.0 to 101% (Iodimetry) |
Linear Formula | Na2S2O3·5H2O |
Merck Index | 15, 8821 |
Solubility Information | Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in turpentine |
Formula Weight | 248.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'