missing translation for 'onlineSavingsMsg'
Learn More
Sodium Thiosulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10294250

Sodium Thiosulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10294250
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10294250

Brand: Fisher Chemical S/7240/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 500g, CHEBI:32150, White

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number 149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrateShow MoreShow Less
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48.5°C
Color White
pH 6.0 to 8.4
Physical Form Solid
Quantity 500 g
Formula Weight 248.18g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Thiosulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.