missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, Extra Pure, SLR, Fisher Chemical™
22.90 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2 kg
3 kg
Specifications
Melting Point | 48.5°C |
Color | White |
pH | 6.0 to 8.4 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 248.18g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'