missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, puriss., Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
4 x 5 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 2.5 kg |
Linear Formula | Na2S2O3 · 5H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 248.18g/mol |
Percent Purity | 99 to 101% |
Grade | puriss. |
Chemical Name or Material | Sodium Thiosulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'