Learn More
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical, Quantity: 1AMP, Packaging: Ampoule, Boiling Point: 100 deg.C, Colorless, MDL Number: 3499, Physical Form: Liquid
Brand: Fisher Chemical J/7950C/05
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7732-18-5 Weight : 0.10000kg
Description
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical, Quantity: 1AMP, Packaging: Ampoule, Boiling Point: 100 deg.C, Colorless, MDL Number: 3499, Physical Form: Liquid
For volumetric analysisSpecifications
Colorless | |
100°C | |
MFCD00003499 | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
1 Ampule |
7772-98-7 | |
Na2O3S2 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate | |
Ampoule | |
Liquid |