missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10753694
5 L, HDPE plastic bottle
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrateShow MoreShow Less
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Liquid
Quantity 5 L
Packaging HDPE plastic bottle

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2525707 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberJ/7950C/05,J/7950/24,J/7950C/90,J/7950/21,J/7950/17,J/7950/27,J/7950/15
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.