missing translation for 'onlineSavingsMsg'
Learn More

Sodium trichloroacetate, 97%

Product Code. 11408856
Click to view available options
Quantity:
10 g
250 g
1000 g
5000 g
Unit Size:
1000g
10g
250g
5000g
This item is not returnable. View return policy

Product Code. 11408856

Brand: Thermo Scientific Alfa Aesar A17004.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 650-51-1 | C2Cl3NaO2 | 185.36 g/mol

Sodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.

Solubility
Soluble in water, ethanol, methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 650-51-1
Molecular Formula C2Cl3NaO2
Molecular Weight (g/mol) 185.36
MDL Number MFCD00064198
InChI Key SAQSTQBVENFSKT-UHFFFAOYSA-M
Synonym sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca
PubChem CID 23681045
ChEBI CHEBI:81976
IUPAC Name sodium;2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 250 g
UN Number UN3077
Beilstein 3572664
Sensitivity Hygroscopic
Merck Index 14,9627
Solubility Information Soluble in water,ethanol,methanol,acetone.
Formula Weight 185.37
Percent Purity 97%
Chemical Name or Material Sodium trichloroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.