missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 10702253
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-30-9
Molecular Formula CF3NaO3S
Molecular Weight (g/mol) 172.05
MDL Number MFCD00061607
InChI Key XGPOMXSYOKFBHS-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
PubChem CID 23676748
IUPAC Name sodium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 253.0°C to 255.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Na)
Packaging Glass bottle
Linear Formula F3COSNa
Quantity 25 g
Solubility Information Solubility in water: soluble in water.
Formula Weight 172.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium trifluoromethanesulfonate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium trifluoromethanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'