Learn More
Sodium trimethylsilanolate, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
£105.00 - £723.80
Chemical Identifiers
CAS | 18027-10-6, 109-99-9 |
---|---|
Molecular Formula | C3H9NaOSi |
Molecular Weight (g/mol) | 112.18 |
MDL Number | MFCD00013926 |
InChI Key | HSNUIYJWTSJUMS-UHFFFAOYSA-N |
Synonym | sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat |
PubChem CID | 23672319 |
IUPAC Name | sodium;trimethyl(oxido)silane |
SMILES | C[Si](C)(C)[O-].[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18027-10-6, 109-99-9 | |
112.18 | |
HSNUIYJWTSJUMS-UHFFFAOYSA-N | |
23672319 | |
C[Si](C)(C)[O-].[Na+] |
C3H9NaOSi | |
MFCD00013926 | |
sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat | |
sodium;trimethyl(oxido)silane |
Specifications
18027-10-6, 109-99-9 | |
MFCD00013926 | |
sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat | |
C[Si](C)(C)[O-].[Na+] | |
112.18 | |
112.18 |
C3H9NaOSi | |
−17°C | |
HSNUIYJWTSJUMS-UHFFFAOYSA-N | |
sodium;trimethyl(oxido)silane | |
23672319 | |
Sodium trimethylsilanolate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 241-939-8
- Sodium trimethylsilanolate, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only