missing translation for 'onlineSavingsMsg'
Learn More

Sodium trimethylsilanolate, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10406060
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18027-10-6, 109-99-9
Molecular Formula C3H9NaOSi
Molecular Weight (g/mol) 112.18
MDL Number MFCD00013926
InChI Key HSNUIYJWTSJUMS-UHFFFAOYSA-N
Synonym sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat
PubChem CID 23672319
IUPAC Name sodium;trimethyl(oxido)silane
SMILES C[Si](C)(C)[O-].[Na+]

Specifications

Quantity 100 mL
Flash Point −17°C
Formula Weight 112.18
Packaging AcroSeal™ Glass Bottle
Chemical Name or Material Sodium trimethylsilanolate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium trimethylsilanolate, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'