missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Triphosphate Pentabasic, purum p.a, ≥98.0% (T), Honeywell™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Specifications
Specifications
Melting Point | 622°C |
Color | White |
pH | 9.5 to 10.3 |
Physical Form | Granules |
Quantity | 500 g |
Linear Formula | Na5P3O10 |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water |
Formula Weight | 367.86g/mol |
Percent Purity | ≥98.0% (T) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'