Learn More
Sodium tripolyphosphate, Thermo Scientific Chemicals
£26.90 - £59.90
Chemical Identifiers
CAS | 7758-29-4 |
---|---|
Molecular Formula | Na5O10P3 |
Molecular Weight (g/mol) | 367.86 |
MDL Number | MFCD00003514 |
InChI Key | HWGNBUXHKFFFIH-UHFFFAOYSA-I |
Synonym | sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n |
PubChem CID | 24455 |
IUPAC Name | pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate |
SMILES | [O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366778
|
Thermo Scientific Alfa Aesar
013440.36 |
500 g |
£26.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376778
|
Thermo Scientific Alfa Aesar
013440.A3 |
2 kg |
£59.90
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tripolyphosphate is used in water softening, heavy-duty fabric washing, automatic dish washing, drilling fluid cleaning and liquid cattle feed supplements. It is also used as a peptizing, emulsifying, dispersing agent and as a hydrogen peroxide solution stabilizer. It acts as a preservative for seafood, meats, poultry, and animal feeds. It finds application as a polyanion crosslinker in polysaccharide based drug delivery. Further, it is used as a sealant for leaking farm ponds and a calcium and magnesium chelating agent. In addition to this, it is used in ceramics, leather tanning, flame retardants, paper pulping, paper coatings, textile processing, ore flotation, anticorrosion pigments and rubber production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-29-4 | |
367.86 | |
HWGNBUXHKFFFIH-UHFFFAOYSA-I | |
24455 | |
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] |
Na5O10P3 | |
MFCD00003514 | |
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n | |
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate |
Specifications
622°C | |
White | |
Na5O10P3 | |
14,8697 | |
Soluble in water; Slightly soluble in methanol; Insoluble in diethyl ether and n-octanol. | |
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] | |
367.86 | |
367.91 | |
Hygroscopic |
7758-29-4 | |
Granules | |
MFCD00003514 | |
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n | |
HWGNBUXHKFFFIH-UHFFFAOYSA-I | |
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate | |
24455 | |
Odorless | |
Sodium tripolyphosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 231-838-7
missing translation for 'rtecsNumber' : YK4570000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium tripolyphosphate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Pentasodium triphosphate
RUO – Research Use Only