missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tungstate dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
£19.90 - £154.77
Chemical Identifiers
CAS | 10213-10-2 |
---|---|
Molecular Formula | Na2O4W·2H2O |
Molecular Weight (g/mol) | 329.85 |
MDL Number | MFCD00149190 |
InChI Key | WPZFLQRLSGVIAA-UHFFFAOYSA-N |
Synonym | sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o |
PubChem CID | 150191 |
ChEBI | CHEBI:63939 |
IUPAC Name | disodium;dioxido(dioxo)tungsten;dihydrate |
SMILES | O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10495471
|
Thermo Scientific Acros
424470050 |
5 g | Glass bottle |
£19.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10093863
|
Thermo Scientific Acros
424471000 |
100 g | Glass bottle |
£45.23
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10055860
|
Thermo Scientific Acros
424475000 |
500 g | Glass bottle |
£154.77
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentIdentificadores de productos químicos
10213-10-2 | |
329.85 | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
150191 | |
disodium;dioxido(dioxo)tungsten;dihydrate |
Na2O4W·2H2O | |
MFCD00149190 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
CHEBI:63939 | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] |
Especificaciones
10213-10-2 | |
Slightly Crystalline Powder or Flakes | |
Na2O4W·2H2O | |
MFCD00149190 | |
15, 8826 | |
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] | |
329.85 | |
CHEBI:63939 | |
≥99% | |
Sodium tungstate dihydrate |
White to Yellow | |
99+% | |
Na2WO4·2H2O | |
15,295 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)tungsten;dihydrate | |
150191 | |
329.85 | |
ACS Reagent |
Seguridad y manipulación
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- Sodium tungstate dihydrate
Palabra de advertencia
- Warning
Clasificación
- Acute toxicity Category 4
Indicaciones de peligro
- H302-Harmful if swallowed.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only