missing translation for 'onlineSavingsMsg'
Learn More
Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10117423

Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10117423
5 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10117423

Brand: Thermo Scientific Acros 207620050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-10-2
Molecular Formula Na2O42H2O
Molecular Weight (g/mol) 329.85
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2oShow MoreShow Less
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Physical Form Crystalline Flakes or Powder
Quantity 5 g
Assay Percent Range 99% min
Linear Formula Na2WO4·2H2O
Fieser 15,295
Merck Index 15,8826
Solubility Information Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids
Formula Weight 329.85
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Density 4.18
Chemical Name or Material Sodium tungstate dihydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460622 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number207620025,207620050,207625000,207621000
Lot NumberA0456668 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number207620025,207620050,207625000,207621000
Lot NumberA0451963 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number207620025,207620050,207625000,207621000
Lot NumberA0449531 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number207620025,207620050,207625000,207621000
Lot NumberA0398319 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number207620025,207620050,207625000,207621000
compliance-icons
Product Identifier
  • Sodium tungstate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium tungstate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.