missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number 10792821
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
10213-10-2
Na2O42H2O
329.85
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
10213-10-2
Na2O42H2O
329.85
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Flakes or Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula Na2WO4·2H2O
Fieser 15,295
Merck Index 15,8826
Solubility Information Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids
Formula Weight 329.85
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Density 4.18
Chemical Name or Material Sodium tungstate dihydrate, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'