missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black/Yellow
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
Brand: Notrax 411C0336BY
Additional Details : Weight : 2.00000kg
Description
- Pebble pattern provides non-directional traction and is easy to clean
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
60 ft. | |
Industrial and Commercial Matting | |
91 cm |
Black/Yellow | |
Microcellular Vinyl | |
1830 cm |