Learn More
Diversey™ Soft Care Des E H5 Hand Disinfectant Gel
Soft Care Des E is an alcohol based (71.5%) hand disinfectant gel for waterless skin disinfection suitable for use in all areas, including healthcare and food processing environments.
Brand: Diversey™ 101105801
Additional Details : CAS Number : 64-17-5 Weight : 4,400.00000kg
Description
- Hand disinfectant based on ethanol, a disinfectant which is very effective against resident and transient flora such as bacteria (e.g. MRSA) and yeast
- Also effective against enveloped viruses, such as influenza and certain non-enveloped viruses such as Norovirus and Adenovirus
- Contains a skin softener helping the skin to remain smooth and intact, particularly in areas where frequent hand disinfection is required
- Perfume-free, eliminating the risk of tainting foods and making it suitable for application in food processing areas
- Skin compatibility dermatologically tested (Patch and TEWL tests)
Specifications
Disinfectant Gel | |
Gel | |
Yes | |
Non-sterile |
Clear | |
Single-use | |
10 Bottles | |
0.5 L |