Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
With slim design to allow the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
£197.00 - £235.00
Specifications
Number of Channels 1
Type Fixed Volume Pipette
Quantity 1/Pk.
View More Specs
Products 9
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
12342018
View Documents
Hamilton™
55019-09
100 μL Violet
£201.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12332018
View Documents
Hamilton™
55019-07
50 μL Sandstone
£235.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12352018
View Documents
Hamilton™
55019-11
200 μL Mustard
£197.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12362018
View Documents
Hamilton™
55019-13
250 μL Burnt Orange
£197.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12312018
View Documents
Hamilton™
55019-03
10 μL Purple
£197.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12372018
View Documents
Hamilton™
55019-15
300 μL Brick Red
£235.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12382018
View Documents
Hamilton™
55019-17
500 μL Olive Green
£222.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12302018
View Documents
Hamilton™
55019-01
5 μL Steel Blue
£197.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12392018
View Documents
Hamilton™
55019-19
1000 μL Sky Blue
£235.00
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
  • Wide range of volumes available
  • Ejection sleeve made from a high strength, chemically resistant polymer
  • Universal calibration key that reduces the need for costly calibration services
  • Calibration takes only minutes to perform and eliminates downtime
  • Tip nozzle ensures a proper fit with universal pipette tips
  • Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Specifications
1
Fixed Volume Pipette
1/Pk.