Läs mer
Läs mer
Syngene™ Dymension 3 Software
Handla allt< titel> Produkter
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Gbox-Chemi-XX6/9
Förpackningsstorlek
Each
Beskrivning
- Software for the rapid analysis of 2D protein spots from multiple gels including multi-stained fluorescent gels (Cy2, Cy3, Cy5)
- With interactive process flow diagram
- Main pane and secondary panes for viewing replicates and samples
- Synchronised zoom with intensity autoscale
- Automatic and precise image warping with vector view
- Manual warp editing
- Automatic background subtraction, noise filtering
- SYPRO Ruby crystal filter
- Automatic spot detection
- Region of interest option
- Gaussian spot modelling option
- 3 x 10MB images spot detected in eight seconds
- Spot detection with user adjustable parameters
- Spot editing, filtering and normalisation
- Spot matching across multiple gels
- Manual match editing pI and MW ladder calibration
- Protein list assignment of spots with known identity, pI and MW
- Histogram (brightness and contrast)
- Main pane and secondary panes for multi-stained gel sets, replicates and samples
- Bar chart 3D spot profile
- Matched spot plot
- Correlation plot
- Table of spot matching results
- Scatter plot
- Statistical tests
- Export to Word and Excel
- Spot picker interface
- Supplied with one years' support
Specifikationer
Specifikationer
| For Use With (Application) | 2d electrophoresis analysis |
| For Use With (Equipment) | Gbox-Chemi-XX6/9 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering