Learn More
Retsch™ Software EasySieve™
Retsch™ Software EasySieve™ is software designed for particle size analysis.
Brand: Retsch™
Additional Details : Weight : 0.26100kg
Description
- Automatic registration, evaluation and administration of measurement data
- Logical design, self-explanatory
- Measurement protocol in accordance with standards
- Easy selection and configuration of common analytical scales
- English and German
- For automatic control of sieve shakers AS 200 control, AS 300 control, AS 450 control, AS 400 control, AS 200 tap, AS 200 jet
- For Windows™ 2000 / XP / Vista / 7 / 8
Specifications
Sieve Shakers AS control / AS 200 tap / AS jet | |
Software | |
Particle Size Analysis |