Learn More
Cole-Parmer™ Soil Auger Kit with Carrying Case
Soil Auger Kit with Carrying Case - Obtain 3 in. diameter samples in nearly all soil conditions with this complete auger kit.
Brand:  Cole-Parmer™ 99026-40
Product Code. 11739128
  • £2520.00 / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Nickel-plated carbon steel material protects against rust and wear
Specifications
Specifications
Nickel-plated
Soil Auger Kit
3 in. general-purpose auger, 3 in. clay auger, 3 in. sand auger, 3 in. mud auger, handgrip, two 45 in. auger extensions, two tools to dissemble extensions, carrying case