Learn More
Brady™ Brass Padlocks
Features solid brass body and shackle that resists corrosion and sparks.
Brand:  Brady™ 051355
 View more versions of this product
Product Code. 11886073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 6-pin cylinder offers more unique key cuts for better tamper resistance
  • Locks are protected by coloured vinyl sheaths making it easy to color-code your lockout program
  • Padlocks are available keyed different
  • Lock labels sold separately
  • Two keys supplied with each padlock
Specifications
Specifications
Brass Padlocks
Blue
Brass Padlocks
43.00 x 60.00 mm
21.00 mm
6
33.5 mm
51355
Brass
6.5 mm
21 mm
Brass
12 Pk.