missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Solid Cap

33 mm threaded cap
Brand: Corning™ 3568
This item is not returnable.
View return policy
Description
(This is the suggested replacement for 3280)
These disposable high density polyethylene sterile solid caps are designed to fit the Corning™ 500 mL, 1 L and 3 L Disposable Spinner Flasks (Cat. Nos. 3153, 3561 and 3563). They also fit Corning disposable polystyrene and polycarbonate storage bottles and glass bottles with GL45 threads.
- 33mm threaded cap
- Vented cap has 0.2 μm pore nonwettable (hydrophobic) membrane sealed directly to the cap
- Allows gas exchange while minimizing the risk of contamination
- Sterilized by gamma irradiation

Specifications
4 | |
Solid Cap | |
Sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Solid Cap > Product Type: GL45
Spot an opportunity for improvement?Share a Content Correction