missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Solid Container with Attached Lid

Solid Container with Attached Lid has a hinge-attached lid to better protect contents with no risk of losing the lid. An ideal container for transporting tableware.

£66.90 - £99.65

Specifications

Type Storage Tote
Material Polypropylene
Color Gray
Shape Rectangular
Closure Type Hinge-attached Lid
View More Specs

Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
17998741
View Documents
Gilac™
G121002
400 x 300 x 330 mm
£66.90
Each
Please to purchase this item. Need a web account? Register with us today!
17978661
View Documents
Gilac™
G121003
600 x 400 x 246 mm
£99.65
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Solid container with hinge-attached lid to better protect contents.
  • No risk of losing the lid.
  • Ideal container for transporting tableware.
  • Designed with smooth sides for easy cleaning.
  • Ergonomic integrated handles on all four sides.
  • Stackable container with vertical reinforcement for greater durability.
  • Compatible with 600 x 400 dollies for easy transport.
  • Easy-to-seal lids with cord to prevent theft.
Specifications

Specifications

Storage Tote
Gray
Hinge-attached Lid
Polypropylene
Rectangular
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilac™ Solid Container with Attached Lid

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.