missing translation for 'onlineSavingsMsg'
Learn More

Solids Concentrate Wastewater QC Sample, SPEX CertiPrep™

Solids Concentrate Wastewater QC Sample, 23 mL vial, SPEX CertiPrep™. ISO Guide 34 Certified

Brand:  SPEX Certiprep 4032

Product Code. 11904974

  • £152.00 / 23mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Wastewater QC Sample
  • 23 mL vial yields 1 L of solution

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Solids Concentrate QC Sample
23 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only