missing translation for 'onlineSavingsMsg'
Learn More

Solids Wastewater QC Sample, Ready to Analyze, SPEX CertiPrep™

Solids QC Samples for Wastewater Analysis, 500 mL, SPEX CertiPrep™

Brand:  SPEX Certiprep 499

Product Code. 11984984

  • £196.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Solids QC Samples
  • Wastewater analysis
  • 500 mL volume

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Solids QC Sample
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Ready to analyze
500 mL
Wastewater Testing
Certified Reference Material
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Solids Wastewater QC Sample, Ready to Analyze, SPEX CertiPrep™ > 500mL; Triple-leached LDPE bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Product intended for laboratory use only