Learn More
3M™ Solus™ Safety Glasses
Product Code. 15749677
Click to view available options
Lens Tint:
Amber
Blue, Mirror
Bronze
Clear
Indoor/Outdoor, Mirror
Red, Mirror
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Unit Size:
Pack of 20
Product Code. 15749677
Brand: 3M™ 7000032478
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Solus™ Safety Glasses are stylish and lightweight for improved comfort and wearer acceptance.
  • Lens curve has been calculated to optimize the fitting, field of vision and coverage of the eyes
  • Extremely stylish and lightweight glasses
  • Soft inner padding of the temple, leading to increased comfort
  • Supplied with a microfiber storage bag for cleaning and protection
  • Anti-scratch and anti-fog coating for increased durability and improved vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN 166: 2001
Lens Coating Scratch-resistant
Lens Tint Blue, Mirror
Product Type Safety glasses
Frame Color Silver, Blue
Includes Microfiber storage bag
Lens Material Polycarbonate