missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solutions A+ Aloe Latex Gloves

Product Code. 15183269
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Protects hands from drying, chapping and cracking. Fisherbrand™ Solutions A+ Aloe Latex Gloves combine protection and comfort.

Latex gloves provide a protective barrier and contain soothing Aloe vera to help prevent skin from drying, cracking and chapping. Textured to provide excellent grip. Powder free. EN 374:2003 (AQL 1.5).
  • PPE Category III
  • Disposable
  • Aloe vera coating protects from drying, chapping and cracking
  • Ideal for prolonged use
  • Textured surface provides good grip
  • Latex
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved
Color Natural
Hand Ambidextrous
Material Latex
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Thickness (English) 5.5 mil
For Use With (Application) Ideal for workers who have frequent glove changes
Glove Cuff Style Beaded/Rolled
Tensile Strength 18 MPa
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.