missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Solutions for perfectION™ Combined ISE

Product Code. p-4676283 Shop All Mettler Toledo Products
Click to view available options
Filling Solution Type:
Electrolyte B
Electrolyte C
Electrolyte D
Electrolyte E
Electrolyte F
Applications:
Chloride, Cyanide, Lead, or Silver/Sulfide
Copper, Iodide
Nitrate
Potassium
Silver
Unit Size:
300mL
This item is not returnable. View return policy

Product Code. 12681296

Brand: METTLER TOLEDO™ 51344755

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended reference solution for replacing electrolyte in ion selective electrodes.

  • Keep electrolyte levels high by refilling regularly - when the probe is submerged, the electrolyte level should always be higher than the sample level.
  • Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
  • Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
TRUSTED_SUSTAINABILITY

Specifications

Applications Nitrate
Filling Solution Type Electrolyte F
Model perfectION ion electrolyte E, 5 x 60 mL
Includes 5 x 60 mL
For Use With (Equipment) perfectION Electrode
Temperature (Metric) Operating 0°C to 60°C
Type Combined ISE Solution
Product Identifier
  • perfectION Ion Electrolyte F
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P202-Do not handle until all safety precautions have been read and understood.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium Sulfate, Silver Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.