Learn More
Cytiva Sonicated DNAs for Hybridization Studies
Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.
Brand: Cytiva 27-4565-01
350.89 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Fragmented DNA with a size range from 200-6000 nucleotides
- Purified by phenol extraction and freeze dried
- Tested for absence of nuclease
- DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp
Specifications
Solid | |
-20°C | |
Hybridisation experiments |
Sonicated Salmon Sperm DNA | |
100/Pk. |