Learn More
Sonoco™ ThermoSafe PolarPack™ MoistureGuard™
Moisture resistance gel pack. Minimize condensation, these packs have a multi-layered exterior, protecting product from moisture damage.
£16.07
Specifications
For Use With (Application) | For shipping temperature critical packages during transit |
---|---|
Freezing Point | 0°C |
Product Code | Brand | Dimensions (L x W x H) | Weight (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Weight (English) | Price | Quantity & Availability | ||||
13591930
|
Sonoco™
MG16 |
6.5 x 5.5 x 0.75 in. | 16 oz. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Outer film layer assures products arrive in top condition by absorbing condensation.
- Provide refrigeration for transport of food, pharmaceuticals, and medical products.
- Condition coolant in freezer before shipments.
- Available in several sizes
Specifications
For shipping temperature critical packages during transit | |
0°C |