missing translation for 'onlineSavingsMsg'
Learn More
Sorbitan monolaurate, Thermo Scientific Chemicals Product Code.: 11338615

Sorbitan monolaurate, Thermo Scientific Chemicals

Product Code. 11338615
250 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11338615

Brand: Thermo Scientific Alfa Aesar L12099.30

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

Solubility
Soluble in 2-ethoxyethanol, ethanol and methanol. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-39-2
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00005365
InChI Key LWZFANDGMFTDAV-IOVMHBDKSA-N
Synonym Sorbitan monododecanoate; Span™ 20
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Density 1.032
Quantity 250 g
Flash Point >110°C (230°F)
Refractive Index 1.474
Merck Index 14,8724
Solubility Information Soluble in 2-ethoxyethanol,ethanol and methanol. Not miscible or difficult to mix in water.
Formula Weight 346.47
Chemical Name or Material Sorbitan monolaurate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242951 Certificate TypeCertificate of Analysis Date24/08/2022 Catalog NumberL12099.30,L12099.0B
Lot Number10174175 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12099.30,L12099.0B
Lot Number10147930 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12099.30,L12099.0B
Lot Number10132433 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12099.30,L12099.0B
Lot Number10194532 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12099.30,L12099.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sorbitan monolaurate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.